Learn More
CD142 Mouse anti-Human, PE, Clone: HTF-1, BD
Mouse Monoclonal Antibody
$170.00 - $332.00
Specifications
Antigen | CD142 |
---|---|
Clone | HTF-1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
Reacts with a 45-47 kDa, single chain, type I transmembrane protein also known as Tissue Factor (TF). CD142 has been referred in the literature as coagulation Factor III or thromboplastin and it is expressed on activated endothelial cells and lipopolysaccharide (LPS)-stimulated monocytes/macrophages. TF associates with factor VIIa to form a complex and acts as an enzyme that initiates the blood coagulation cascade. It is known as the major initiator of clotting in normal hemostasis. CD142 can be induced by various inflammatory mediators in monocytes and vascular endothelial cells. This antibody is useful in inflammation, thrombosis and hemostasis research.
Specifications
CD142 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HTF-1 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.