Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD143 Mouse anti-Human, Brilliant Violet 711, Clone: BB9, BD Optibuild™

Catalog No. BDB742899 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742899 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742899 Supplier BD Biosciences Supplier No. 742899
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The BB9 monoclonal antibody specifically binds Angiotensin-converting enzyme (ACE) which is also known as Dipeptidyl carboxypeptidase 1 (DCP1) or CD143. CD143 is a type I transmembrane glycoprotein that functions as a zinc metallopeptidase involved with the metabolism of angiotensin, bradykinin, and other oligopeptides. It is expressed on primitive hematopoietic progenitor cells (HPC). BB9 reacts with stromal cells, a minor subset of human bone marrow cells; it does not react with peripheral blood leucocytes. BB9 recognizes a subset of CD34+CD90+ cells that demonstrate low or absent expression of CD38, low retention of Rhodamine 123 and are Ki-67-negative. CD34+BB9+ cells are able to sustain hematopoiesis in pre-CFU culture stimulated with IL-3, IL-6, G-CSF, and SCF. BB9 also reacts with a subset of CD34+ cells from mobilized peripheral blood. This antibody was generated using human bone marrow-derived stromal cells as immunogen.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD143
Classification Monoclonal
Clone BB9
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ACE1, DCP, DCP1, Kininase II, ICH, MVCD3, peptidase P
Host Species Mouse
Immunogen Human bone marrow-derived stromal cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.