Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD146 Monoclonal Antibody (P1H12), NovaFluor™ Blue 610-70S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
Antigen | CD146 |
---|---|
Clone | P1H12 |
Concentration | 0.2 μg/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H058T03B06A
![]() |
Invitrogen™
H058T03B06A |
100 Tests |
Each for $430.00
|
|
|||||
H058T02B06A
![]() |
Invitrogen™
H058T02B06A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD146 Monoclonal antibody specifically detects CD146 in Canine, Human, Mouse, Rabbit samples. It is validated for Flow CytometrySpecifications
CD146 | |
0.2 μg/Test | |
Monoclonal | |
Mouse | |
Human, Mouse, Canine, Rabbit | |
P43121, Q8R2Y2 | |
100342848, 4162, 489368, 84004 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
P1H12 | |
Flow Cytometry | |
NovaFluor Blue 610-70S | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
1-gicerin; AV025631; CD146; CD149; Cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; HEMCAM; l-gicerin; l-gicerin precursor; l-gicerin protein; LOW QUALITY PROTEIN: cell surface glycoprotein MUC18; MCAM; melanoma adhesion molecule; melanoma cell adhesion molecule; Melanoma-associated antigen A32; melanoma-associated antigen MUC18; Muc18; mucin-18; s-endo; S-endo 1 endothelial-associated antigen; s-gicerin; s-gicerin precursor | |
IgG1 κ | |
Affinity chromatography | |
MCAM |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title