Learn More
Description
Specifications
Specifications
| Antigen | CD146 |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone P1H12 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | MCAM; MELCAM; MUC18; Gicerin; Melanoma cell adhesion molecule |
| Host Species | Mouse |
| Immunogen | Human Umbilical Vein Cells |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.