Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD146 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501567459
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7459 Supplier Bethyl Laboratories, Inc Supplier No. A304335A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CD146 (MCAM) is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. CD146 is expressed at cell-cell junctions in all endothelial cells, and has also been observed in other normal and maligt cell types. In addition to its recognized role as a calcium-independent cell adhesion molecule, CD146 may also act as a signal transduction molecule in the recruitment of Fyn kinase, and in the subsequent tyrosine-phosphorylation of intracellular proteins involved in actin cytoskeleton assembly. CD146 plays a role in cell adhesion, and in cohesion of the endothelial monolayer at intercellular junctions in vascular tissue. CD146 expression may allow melanoma cells to interact with cellular elements of the vascular system, thereby enhancing hematogeneous tumor spread. Further, CD146 could be an adhesion molecule active in neural crest cells during embryonic development. CD146 acts also as surface receptor that triggers tyrosine phosphorylation of FYN and PTK2, and a transient increase in the intracellular calcium concentration.

Specifications

Antigen CD146
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MCAM
Gene Accession No. P43121, Q8R2Y2
Gene Alias CD146, MUC18, MCAM, Gicerin, S-endo 1 endothelial-associated antigen, cell surface glycoprotein MUC18, cell surface glycoprotein P1H12, melanoma adhesion molecule, melanoma-associated antigen A32, melanoma-associated antigen MUC18
Gene Symbols Mcam
Host Species Rabbit
Immunogen Between 600 and 650
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4162, 84004
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.