Learn More
CD147 Rat anti-Mouse, BUV496, Clone: RL73 (also known as RL73.2), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD147 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | RL73 (also known as RL73.2) |
| Conjugate | Brilliant Ultraviolet 496 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Bsg; Basigin; BASI; EMMPRIN; gp 42; HT7; HT-7; Neurothelin |
| Host Species | Rat |
| Immunogen | Mouse EL4 Cell Subline |
| Quantity | 50 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.