Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD147 Recombinant Rabbit Monoclonal Antibody (125)
GREENER_CHOICE

Catalog No. PIMA529060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529060 Supplier Invitrogen™ Supplier No. MA529060
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human CD147/Basigin/BSG.

CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and malignant tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD147
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone 125
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 10% trehalose and no preservative; pH 7.0
Gene BSG
Gene Accession No. P35613
Gene Alias 5A11; 5A11 antigen; 5A11/Basigin; 5A11/Basigin-2; 5F7; AI115436; AI325119; Basic immunoglobulin superfamily; basigin; basigin (Ok blood group); Basigin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); basignin; Basignin (Ox47 antigen or CE-9) (EMMPRIN in human) (neurothelin HT7 or 5A11 in avian); blood-brain barrier HT7 antigen; BSG; CD147; CD147 antigen; CE9; Collagenase stimulatory factor; EMMPRIN; Extracellular matrix metalloproteinase inducer; glycoprotein CE9; gp 42; gp42/basigin/OX-47/HT7; HT7; HT-7; HT7 antigen; immunoglobulin gene superfamily member; leukocyte activation antigen M6; Membrane glycoprotein gp42; neurothelin; OK; OK blood group antigen; OX-47 antigen; Ox47R; RPE7; TCSF; tumor cell-derived collagenase stimulatory factor; Unknown (protein for MGC:128392); unnamed protein product; UNQ6505/PRO21383
Gene Symbols BSG
Host Species Rabbit
Immunogen Recombinant Human CD147 protein (Met1-His205).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.