Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD148 Hamster anti-Mouse, Brilliant Violet 510, Clone: 8A-1, BD Optibuild™

Catalog No. BDB743843 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743843 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743843 Supplier BD Biosciences Supplier No. 743843
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

The 8A-1 monoclonal antibody specifically binds to CD148, which is also known as Protein tyrosine phosphatase receptor type J (Ptprj), Receptor-type tyrosine-protein phosphatase eta (R-PTP-eta, PTPη ), or Density-enhanced phosphatase-1 (DEP-1). CD148 is a receptor-like tyrosine phosphatase that plays important roles in regulating cell growth, proliferation, differentiation, adhesion, and migration. It is expressed on a wide variety of cell types including epithelial cells, endothelial cells, and fibroblasts. For cells of hemopoietic origin, it is variably expressed on macrophages, neutrophils, dendritic cells, and on developing and mature B cells. CD148 is expressed on platelets and can regulate platelet activation. Although expressed at low levels on thymocytes and mature T cells, its expression is upregulated upon T cell receptor (TCR)-mediated stimulation and may play a role in regulating TCR signaling. CD148, along with CD45, are critical protein tyrosine phosphatases for regulating Src family kinase signaling networks in immune cells.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD148
Classification Monoclonal
Clone 8A-1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ptprj, R-PTP-J, BET, Byp, DEP-1, PTPbeta2, Ptpb2, R-PTP-eta, Scc-1, Scc1
Host Species Syrian Hamster
Immunogen Mouse CD148 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.