Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD148 Hamster anti-Mouse, BUV395, Clone: 8A-1, BD Optibuild™

Catalog No. BDB743848 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743848 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743848 Supplier BD Biosciences Supplier No. 743848
Add to Cart
Add to Cart

Syrian Hamster Monoclonal Antibody

The 8A-1 monoclonal antibody specifically binds to CD148, which is also known as Protein tyrosine phosphatase receptor type J (Ptprj), Receptor-type tyrosine-protein phosphatase eta (R-PTP-eta, PTPη ), or Density-enhanced phosphatase-1 (DEP-1). CD148 is a receptor-like tyrosine phosphatase that plays important roles in regulating cell growth, proliferation, differentiation, adhesion, and migration. It is expressed on a wide variety of cell types including epithelial cells, endothelial cells, and fibroblasts. For cells of hemopoietic origin, it is variably expressed on macrophages, neutrophils, dendritic cells, and on developing and mature B cells. CD148 is expressed on platelets and can regulate platelet activation. Although expressed at low levels on thymocytes and mature T cells, its expression is upregulated upon T cell receptor (TCR)-mediated stimulation and may play a role in regulating TCR signaling. CD148, along with CD45, are critical protein tyrosine phosphatases for regulating Src family kinase signaling networks in immune cells.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD148
Classification Monoclonal
Clone 8A-1
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ptprj, R-PTP-J, BET, Byp, DEP-1, PTPbeta2, Ptpb2, R-PTP-eta, Scc-1, Scc1
Host Species Syrian Hamster
Immunogen Mouse CD148 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.