Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD15/Lewis X Rabbit anti-Human, Clone: BRA-4F1, Novus Biologicals™

Rabbit Monoclonal Antibody
$269.00 - $584.50
Specifications
Antigen | CD15/Lewis X |
---|---|
Clone | BRA-4F1 |
Dilution | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
CD15/Lewis X Monoclonal antibody specifically detects CD15/Lewis X in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
CD15/Lewis X | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
RUO | |
PBS | |
2526 | |
IgG κ | |
Protein A purified |
BRA-4F1 | |
Monoclonal | |
Rabbit | |
Human | |
CD 15, CD-15, Lewis X, Lewis-X, LeX, SSEA-1, stage-specific embryonic antigen 1 | |
This antibody was raised by immunzing mice with human CD15/Lewis X. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title