Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUT4 Monoclonal Antibody (CL6334)
GREENER_CHOICE

Catalog No. PIMA531432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531432 Supplier Invitrogen™ Supplier No. MA531432
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: RKPSGAGWEK EWAEAPQEA Highest antigen sequence indentity to the following orthologs: Mouse - 0%, Rat - 0%.

FUT4 (Fucosyltransferase 4) is a Protein Coding gene. Diseases associated with FUT4 include Colon Adenocarcinoma and Liver Lymphoma. Among its related pathways are Glycosphingolipid biosynthesis - lacto and neolacto series and Mannose type O-glycan biosynthesis. Gene Ontology (GO) annotations related to this gene include fucosyltransferase activity and alpha-(1->3)-fucosyltransferase activity. The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUT4
Applications Western Blot
Classification Monoclonal
Clone CL6334
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUT4
Gene Accession No. P22083
Gene Alias 3-fucosyl-N-acetyl-lactosamine epitope; AI451562; alpha (1,3) fucosyltransferase, myeloid specific; alpha (1,3) fucosyltransferase, myeloid-specific; alpha 1,3-fucosyltransferase; alpha 1,3-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha 13-fucosyltransferase Fuc-T (similar to mouse Fut4); alpha-(1,3)-fucosyltransferase 4; CD15; ELAM ligand fucosyltransferase; ELAM-1 ligand fucosyltransferase; ELFT; FAL; FCT3A; fucosyltransferase 4; fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific); fucosyltransferase IV; Fuct; FUC-TIV; FucT-IV; FUT4; FUTIV; Galactoside 3-L-fucosyltransferase; Lewis x; LeX; Ssea1; SSEA-1; stage-specific embryonic antigen 1
Gene Symbols FUT4
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human FUT4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.