Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD15 Monoclonal Antibody (MMA)
GREENER_CHOICE

Catalog No. PIMA126893
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA126893 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA126893 Supplier Invitrogen™ Supplier No. MA126893
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: Tonsil..

CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MMA
Conjugate Unconjugated
Formulation PBS with 0.09% Sodium azide
Gene FUT4
Gene Accession No. P22083, Q11127
Gene Alias FUTIV; Lewis X; SSEA-1
Gene Symbols FUT4
Host Species Mouse
Immunogen U937 histiocytic cell line.
Purification Method IgM
Quantity 6 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14345, 2526
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.