Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD15 Mouse anti-Human, BV 510, Clone: 7C3.rMAb (also known as PMN7C3 or 7C3), BD Horizon™

Catalog No. BDB567961 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567961 25 Tests
BDB567960 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB567961 Supplier BD Biosciences Supplier No. 567961
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD15
Applications Flow Cytometry
Classification Monoclonal
Clone 7C3.rMAb (also known as PMN7C3 or 7C3)
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias LeX; Lewis X; SSEA-1
Host Species Mouse
Immunogen Human Polymorphonuclear Leucocytes
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.