Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD15 Mouse anti-Human, BV 605, Clone: 7C3.rMAb (also known as PMN7C3 or 7C3), BD Horizon™
Mouse Monoclonal Antibody
$232.00 - $454.00
Specifications
Antigen | CD15 |
---|---|
Clone | 7C3.rMAb (also known as PMN7C3 or 7C3) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Violet 605 |
Description
CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow CytometrySpecifications
CD15 | |
Flow Cytometry | |
Brilliant Violet 605 | |
Mouse | |
Human | |
LeX; Lewis X; SSEA-1 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
7C3.rMAb (also known as PMN7C3 or 7C3) | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Polymorphonuclear Leucocytes | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title