Learn More
CD15 Mouse anti-Human , BV421, Clone: HI98 (also known as HIM1), BD Biosciences™
Shop All BD Biosciences ProductsDescription
Specifications
Specifications
| Antigen | CD15 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HI98 (also known as HIM1) |
| Conjugate | Brilliant Violet 421 |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | Lewis X; Le-X; X-Hapten; SSEA-1; 3-FAL |
| Host Species | Mouse |
| Immunogen | Human Mononuclear Cells |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.