Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ CD15, Mouse Monoclonal Antibody, 200μg/mL; Unlabeled; Purified with BSA and azide

Catalog No. MS10338P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS10338P1 500 μL
MS10338P0 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MS10338P1 Supplier Epredia™ Supplier No. MS10338P1

Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD15, Mouse Monoclonal Antibody.

CD15 or X-hapten plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. CD15 is also expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Host Species: Mouse

Clone: SPM490

Isotype: IgM

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: K562 (an erythroid-myeloid leukemia cell line)

Molecular Weight: 220kDa

Positive Control: Reed-Sternberg's cells in Hodgkin's lymphoma

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD15
Applications Immunohistochemistry
Classification Monoclonal
Clone SPM490
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen K562 (an erythroid-myeloid leukemia cell line)
Purification Method Reed-Sternberg's cells in Hodgkin's lymphoma
Quantity 500 μL
Primary or Secondary Primary
Target Species Human
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.