Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Antibody (11G5a) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $501.50
Specifications
Antigen | CD151 |
---|---|
Clone | 11G5a |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | ELISA, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017649
![]() |
Novus Biologicals
NB100648100.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064810
![]() |
Novus Biologicals
NB10064810 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CD151 | |
1.0 mg/mL | |
ELISA, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
CD151 | |
IgG1 | |
Protein G purified |
11G5a | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
P48509 | |
977 | |
Made to Human CD151 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title