Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Antibody (455807), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4609J
Description
CD151 Monoclonal antibody specifically detects CD151 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CD151 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
977 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Immunocytochemistry, CyTOF | |
455807 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
HEK293 human embryonic kidney cell line transfected with mouse CD151, Met1-Tyr253, Accession # BAA22447 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction