Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Antibody (MM0160-6F40), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212144FR
Description
CD151 Monoclonal specifically detects CD151 in Human samples. It is validated for Flow Cytometry.Specifications
CD151 | |
Monoclonal | |
DyLight 680 | |
BAA06229 | |
CD151 | |
Human recombinant CD151 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0160-6F40 | |
Flow Cytometry | |
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
Mouse | |
Protein G purified | |
RUO | |
977 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction