Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD151 Mouse anti-Human, Clone: B-E31, Novus Biologicals™

Mouse Monoclonal Antibody
$377.50 - $722.00
Specifications
Antigen | CD151 |
---|---|
Clone | B-E31 |
Concentration | 1 mg/ml |
Dilution | Flow Cytometry |
Applications | Flow Cytometry |
Description
CD151 Monoclonal antibody specifically detects CD151 in Human samples. It is validated for Flow CytometrySpecifications
CD151 | |
1 mg/ml | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, tetraspanin-24, tspan-24, TSPAN24hemidesmosomal tetraspanin CD151 | |
Eahy 926 cell line. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cell | |
Primary | |
Store at 4C short term. Aliquot and store at -70C long term. Avoid freeze-thaw cycles. |
B-E31 | |
Flow Cytometry | |
Monoclonal | |
Purified | |
RUO | |
PBS. Sterile-filtered through 0.22 um. | |
977 | |
IgG1 κ | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title