Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD152 (CTLA-4) Armenian Hamster anti-Mouse, PE-Cy7, Clone: UC10-4F10-11, MilliporeSigma™

Catalog No. MABF1586 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1586 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1586 Supplier MilliporeSigma Supplier No. MABF1586
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

This armenian hamster monoclonal Anti-CD152 (CTLA-4) Antibody (mouse), PE-Cy7, clone UC10-4F10-11 for the detection of CD152 (CTLA-4).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD152 (CTLA-4)
Applications Flow Cytometry
Classification Monoclonal
Clone UC10-4F10-11
Concentration Please refer to lot specific datasheet.
Conjugate PE-Cyanine7
Gene Symbols Ctla4,Cd152
Host Species Armenian Hamster
Immunogen Immunogen information for this antibody is proprietary information.
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.