Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD152 Mouse anti-Human, Rhesus, Cynomolgus, Baboon, BB515, Clone: BNI3, BD Biosciences™
Mouse Monoclonal Antibody
$220.00 - $454.00
Specifications
| Antigen | CD152 |
|---|---|
| Clone | BNI3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
Description
CD152 Monoclonal antibody specifically detects CD152 in Human, Rhesus, Cynomolgus, Baboon samples. It is validated for Flow CytometrySpecifications
| CD152 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human CTLA4 Recombinant Protein | |
| Primary |
| BNI3 | |
| Monoclonal | |
| Mouse | |
| Human, Rhesus Monkey, Cynomolgus, Baboon | |
| CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title