Learn More
Description
Specifications
Specifications
| Antigen | CD154 |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone MR1 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | CD40 Ligand; CD40L; gp39; Ly-62; Tnfsf5; T-BAM; HIGM1; IMD3 |
| Host Species | Armenian Hamster |
| Immunogen | Activated mouse Th1 clone D1.6 |
| Quantity | 25 Tests |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.