Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD154 (CD40 Ligand) Monoclonal Antibody (MR1), eFluor™ 450, eBioscience™
GREENER_CHOICE

Catalog No. 48154182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-154-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-154-182 Supplier Invitrogen™ Supplier No. 48154182
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Armenian Hamster Monoclonal Antibody

Description: The MR1 monoclonal antibody reacts with mouse CD154, a 39 kDa transmembrane glycoprotein also known as CD40 ligand (CD40L). CD154 is expressed transiently by activated T cells. Through its binding to CD40 on antigen presenting cells (APC) including B cells, monocytes/macrophages, and dendritic cells, it serves a crucial function in T cell-APC cognate interaction. CD154-interaction with CD40 transduces signals for T-dependent B cell activation and induces B cells to enter the cell cycle. For staining for flow cytometric analysis, it is important to stimulate enriched T cells or enriched CD4 cells (using depletion strategy) prior to staining with MR1. Applications Reported: This MR1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR1 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emissi...

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154 (CD40 Ligand)
Applications Flow Cytometry
Classification Monoclonal
Clone MR1
Concentration 0.2 mg/mL
Conjugate eFluor 450
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD40LG
Gene Accession No. P27548
Gene Alias CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Gene Symbols CD40LG
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21947
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
I am unable to analyze my cells stained with eFluor Organic Dyes today. What options do I have?

Our options will depend on the samples you are analyzing.

If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.

For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

What is the difference between eFluor Organic Dyes and eVolve Qdots?

The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.