Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD154 (CD40 Ligand) Recombinant Rat Monoclonal Antibody (YMF323.6)
GREENER_CHOICE

Catalog No. PIMA548082
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548082 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548082 Supplier Invitrogen™ Supplier No. MA548082
Add to Cart
Add to Cart

Rat Recombinant Monoclonal Antibody

CD154 (CD40 Ligand) Recombinant Monoclonal Antibody for Western Blot, Flow

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD154 (CD40 Ligand)
Applications Flow Cytometry, Western Blot
Classification Recombinant Monoclonal
Clone YMF323.6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene CD40LG
Gene Accession No. P29965
Gene Alias CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5
Gene Symbols CD40LG
Host Species Rat
Immunogen Proprietary sequence.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.