Learn More
CD154 Mouse anti-Human, FITC, Clone: TRAP1, BD
Mouse Monoclonal Antibody
$333.50
Specifications
Antigen | CD154 |
---|---|
Clone | TRAP1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Description
The TRAP1 monoclonal antibody specifically binds to CD154. CD154 is a 39 kDa type II membrane glycoprotein that is a member of the tumor necrosis factor superfamily, Tumor necrosis factor ligand superfamily member 5 (TNFSF5). CD154 is expressed on a variety of cell types including activated CD4+ T cells and some CD8+ T cells, NK cells, mast cells and basophils. CD154 is also known as CD40 ligand (CD40L); it serves as a ligand for CD40 that is expressed on B cells, macrophages, and dendritic cells. The expression of CD154 by activated T-helper cells costimulates B-cell activation and proliferation through binding to CD40 expressed on B cells. In response to T-dependent antigens, the CD154 and CD40 interaction is required for B-lymphocyte differentiation, including immunoglobulin production and isotype switching and memory B cell generation. The TRAP1 antibody can partially block T cell-B cell interaction and inhibit the subsequent proliferation, differentiation, and memory formation of B cells. It has been reported that patients with X-linked hyper-IgM syndrome have defective expression of functional CD154 due to mutations in the CD40LG gene that encodes CD154.
Specifications
CD154 | |
Flow Cytometry | |
FITC | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ | |
Affinity Purified | |
CD40LG; CD40 ligand; CD40L; TNFSF5; T-BAM; TRAP |
TRAP1 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.