Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD155 Monoclonal Antibody (3F1)

Catalog No. PIMA533379
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533379 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533379 Supplier Invitrogen™ Supplier No. MA533379

Rat Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL). Application Notes: Flow cytometry - This antibody stains the extracellular domain of mouse CD155. The thymocytes were blocked with 5% mouse serum before incubation with clone 3F1 for 1 hour at 4°C As positive control CD4+CD8+ thymocytes was used and as negative control CD155-/- derived cells. Functional Assay- This antibody functions as antagonist. The antibody was functionally tested by blocking adhesion of Sf9 cells expressing mCD155 to RMA, CD4+ and CD8+ cells. Furthermore, antibody clone 3F1 prevents formation of mCD155/ligand complexes. Positive control: CD4+CD8+ mouse thymocytes; Negative control: CD155-/- cells.

The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD155
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 3F1
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene PVR
Gene Accession No. 0
Gene Alias 3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4
Gene Symbols PVR
Host Species Rat
Immunogen Ectodomain of recombinant His-tagged CD155.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52118
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.