Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD155 Mouse, anti-Mouse, R718, Clone: 3F1, BD OptiBuild™

Catalog No. BDB752114 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752114 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752114 Supplier BD Biosciences Supplier No. 752114
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 3F1 monoclonal antibody specifically binds to CD155, which is also known as Poliovirus receptor (Pvr) or Tumor-associated antigen 1 (Taa1). CD155 is a type I transmembrane glycoprotein that belongs to the Ig supergene family. CD155 is an adhesion receptor that binds to different ligands including nectin-3, CD96, CD226, TIGIT, and the extracellular matrix protein vitronectin. It is highly expressed on double positive thymocytes and variably expressed on mature thymocytes and T cells, including regulatory T cells and NKT cells. CD155 is also differentially expressed on subsets of B cells, plasma cells, dendritic cells, and monocytes. CD155 expression is upregulated by activated T cells, B cells, and dendritic cells. CD155 is involved in forming adherens junctions between adjacent epithelial or endothelial cells. CD155 plays roles in regulating cell growth, adhesion, motility, migration, and cell-mediated cytotoxicity. CD155-deficient mice exhibit impaired secondary humoral immune responses to orally administered antigens.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD155
Applications Flow Cytometry
Classification Monoclonal
Clone 3F1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.