Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD155 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596414
Description
Positive Samples: LO2; Cellular Location: Cell membrane, Single-pass type I membrane protein Immunogen sequence: TCKVEHESFE KPQLLTVNLT VYYPPEVSIS GYDNNWYLGQ NEATLTCDAR SNPEPTGYNW STTMGPLPPF AVAQGAQLLI RPVDKPINTT LICNVTNALG ARQAELTVQV KEGPPSEHSG MSRNAI.
The protein encoded by this gene is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The external domain mediates cell attachment to the extracellular matrix molecule vitronectin, while its intracellular domain interacts with the dynein light chain Tctex-1/DYNLT1. The gene is specific to the primate lineage, and serves as a cellular receptor for poliovirus in the first step of poliovirus replication. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
CD155 | |
Polyclonal | |
Unconjugated | |
PVR | |
3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
52118, 5817 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P15151 | |
PVR | |
Recombinant fusion protein containing a sequence corresponding to amino acids 220-345 of human CD155/PVR (NP_0064964?). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction