Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Antibody (114), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289272AF594
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot, ELISA.Specifications
CD155/PVR | |
Monoclonal | |
Alexa Fluor 594 | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD155/PVR extracellular domain (Met 1-Asn 343; NP_006496.2). | |
0.1 mL | |
Virology Bacteria and Parasites | |
5817 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
114 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction