Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Antibody (19C12), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CD155/PVR |
---|---|
Clone | 19C12 |
Dilution | Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
CD155/PVR | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
A synthesized peptide derived from Human CD155/PVR [UniProt P15151] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
19C12 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Virology Bacteria and Parasites | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
5817 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title