Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Antibody (300907), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB25301PEATT594
Description
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Flow CytometrySpecifications
CD155/PVR | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
300907 | |
Flow Cytometry | |
CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
Mouse myeloma cell line NS0-derived recombinant human CD155/PVR, Gly27-Asn343, Accession # AAH15542 | |
0.1 mL | |
Virology Bacteria and Parasites | |
5817 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction