Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Antibody (BLR074G), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP278711
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD155/PVR | |
Monoclonal | |
Unconjugated | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA. with 0.09% Sodium Azide | |
PVR | |
A recombinant protein representing the extracellular domain of human PVR/CD155. | |
100 μL | |
Primary | |
Human | |
Supernatant |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
BLR074G | |
Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
CD155, CD155 antigen, HVED, Necl-5, NECL5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
Virology Bacteria and Parasites | |
5817 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction