Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD155/PVR Rabbit anti-Mouse, Clone: 305, Novus Biologicals™

Rabbit Monoclonal Antibody
$390.00 - $560.50
Specifications
Antigen | CD155/PVR |
---|---|
Clone | 305 |
Dilution | Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:1000 |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Description
CD155/PVR Monoclonal specifically detects CD155/PVR in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD155/PVR | |
Flow Cytometry 1:25-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
Recombinant Mouse CD155/PVR protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
305 | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Virology Bacteria and Parasites | |
0.2 um filtered solution in PBS | |
5817 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title