Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD156c Mouse anti-Human, Brilliant Violet 786, Clone: 11G2, BD Optibuild™

Catalog No. BDB742792 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742792 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742792 Supplier BD Biosciences Supplier No. 742792
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11G2 monoclonal antibody specifically binds to CD156c which is also known as Disintegrin and metalloproteinase domain-containing protein 10 (ADAM10). CD156c is a ñ70 kDa type I transmembrane glycoprotein that belongs to the ADAM family. It is widely expressed on hematopoietic and non-hematopoietic cells. CD156c is expressed on the plasma membrane, as well as by membranes within intracellular compartments and exosomes. CD156c serves as a broadly-reactive endopeptidase that cleaves membrane-bound proteins in a process known as ectodomain shedding or release. It can cleave transmembrane molecules into soluble forms and thereby regulate the functions of a variety of different receptors and ligands that are involved in cellular signaling and adhesion. These include membrane TNF, CX3CL1, CXCL16, Ephrin-A2, Notch and Delta molecules, cadherins, CD23, CD44, IL-6 Receptor/CD126, CD171, and amyloid precursor protein. CD156c expression may be constitutive or induced, such as in inflamed tissues, including arthritic joints and in the nervous system. Dysregulated CD156c expression may be associated with certain cancers and Alzheimer’s disease.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD156c
Classification Monoclonal
Clone 11G2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ADAM 10, AD10, ADA10, MADM, KUZ, Kuzbanian protein homolog, HsT18717, RAK
Host Species Mouse
Immunogen Human Jurkat Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.