Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD157 Monoclonal Antibody (eBioSY11B5 (SY11B5)), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™

Catalog No. H041T03B08A
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H041T03B08A 100 Tests
H041T02B08A 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H041T03B08A Supplier Invitrogen™ Supplier No. H041T03B08A
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD157, also known as Bone Marrow Stromal Cell Antigen 1 (BST1), is a pleiotropic ectoenzyme and glycosylphosphatidylinositol-anchored molecule that functions both as an enzyme and a receptor. Similar to CD38, CD157 is involved in the metabolism of NAD+, which may play a role in regulating intracellular Ca2+ levels. As an enzyme, CD157 is also referred to as ADP-ribosyl cyclase 2 and DAMP-1, and it facilitates pre-B cell growth. As a receptor, CD157 is thought to initiate a signal transduction cascade upon binding to its putative ligand, leading to the phosphorylation of cytoplasmic proteins, including focal adhesion kinase (FAK). While the mechanism and functional significance of CD157-initiated signal transduction are not fully characterized, its role in cellular signaling is evident. BST1 expression is enhanced in bone marrow stromal cell lines derived from patients with rheumatoid arthritis, suggesting a link to polyclonal B cell abnormalities observed in the disease. This overexpression in the stromal cell population may contribute to the pathophysiology of rheumatoid arthritis, highlighting the importance of CD157/BST1 in both normal immune function and disease states.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD157
Applications Flow Cytometry
Classification Monoclonal
Clone eBioSY11B5 (SY11B5)
Concentration 0.6 μg/Test
Conjugate NovaFluor Blue 660-120S
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Bst1
Gene Accession No. Q10588
Gene Alias 114/A10; A530073F09; ADP-ribosyl cyclase 2; ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 2; antigen BP3; Bone marrow stromal antigen 1; bone marrow stromal cell antigen 1; Bp3; Bp-3; BP-3 alloantigen; Bst1; BST-1; Bsta1; cADPr hydrolase 2; CD157; CD157 antigen; Cyclic ADP-ribose hydrolase 2; leukocyte antigen 65; Ly65; Ly-65; NAD(+) nucleosidase; NAD+ nucleosidase
Gene Symbols Bst1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 683
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.