Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD157 Mouse anti-Human, Unconjugated, Clone: SY/11B5, BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569643
Description
CD157 Monoclonal antibody specifically detects CD157 in Human samples. It is validated for Flow cytometry
Specifications
CD157 | |
Monoclonal | |
0.5 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
683 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Store undiluted at 4°C. | |
IgG1 κ |
Flow Cytometry | |
SY/11B5 | |
Unconjugated | |
BST1; Bone marrow stromal antigen 1; ADP-ribosyl cyclase 2; cADPr hydrolase | |
Human CD157 Transfected Cell Line | |
0.1 mg | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction