Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158a Monoclonal Antibody (HP-3E4), Abbomax

Catalog No. 502204321
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4321 Supplier Abbomax Inc Supplier No. 605140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Tonsil Cellular location: Membrane.

CD158a Monoclonal antibody specifically detects CD158a in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)

Specifications

Antigen CD158a
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HP-3E4
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene KIR2DL1
Gene Accession No. P43626
Gene Alias CD158 antigen-like family member A; CD158a; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 1; killer cell immunoglobulin-like receptor 2DL1; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1; killer Ig receptor; killer inhibitory receptor 2-2-1; KIR221; KIR2DL1; KIR2DS1; KIR-K64; MHC class I NK cell receptor; natural killer cell receptor; Natural killer-associated transcript 1; NKAT; NKAT1; NKAT-1; p58 killer cell inhibitory receptor KIR-K64; p58 natural killer cell receptor clones CL-42/47.11; p58 NK cell inhibitory receptor NKR-K6; p58 NK receptor CL-42/47.11; p58.1; p58.1 MHC class-I-specific NK receptor
Host Species Mouse
Immunogen A recombinant protein of human CD158a
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.