Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158b Mouse anti-Human, Brilliant Violet 510, Clone: CH-L, BD Optibuild™

Catalog No. BDB743452 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743452 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743452 Supplier BD Biosciences Supplier No. 743452
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CH-L monoclonal antibody specifically binds to CD158b proteins. These proteins are 50-58 kDa type I glycoproteins that belong to the Killer cell immunoglobulin-like receptor (KIR) family: (KIR2DL2/L3/S2). They are also known as CD158b1 (KIR2DL2; NKAT-6; p58.2), CD158b2 (KIR2DL3; NKAT-2; p58.2), or CD158j (KIR2DS2; NKAT-5; p50.2). The CD158b molecules are composed of two extracellular Ig-like domains, and a transmembrane region. CD158b1 and CD158b2 also possess long (84 or 76 amino acids, respectively) cytoplasmic tailswith two immunoreceptor tyrosine-based inhibition motifs (ITIM) whereas CD158j has a short (39 amino acid) cytoplasmic tail that lacks the ITIM motif. CD158b molecules are expressed on NK cells and subsets of TCR αβ+ cells or TCR γδ+ cells. Ligand- or CH-L antibody-bound CD158b1 or CD158b2 can reportedly inhibit cytolytic NK and T cell responses to various stimuli including certain target cells expressing MHC class I ligands encoded by HLA-C alleles (Cw 1, 3, 7 and 8). CD158j reportedly can enhance some cellular cytolytic responses.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158b
Classification Monoclonal
Clone CH-L
Concentration 0.2mg/mL
Conjugate Brilliant Violet 510
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD158b1/KIR2DL2/NKAT-6, CD158b2/KIR2DL3/NKAT-2, CD158j/KIR2DS2/NKAT-5
Host Species Mouse
Immunogen Human NK Cells
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.