Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD158b Mouse anti-Human, RY586, Clone: CH-L, BD OptiBuild™

Catalog No. BDB753536 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB753536 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB753536 Supplier BD Biosciences Supplier No. 753536
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD158b Monoclonal antibody specifically detects CD158b. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD158b
Applications Flow Cytometry
Classification Monoclonal
Clone CH-L
Concentration 0.2 mg/mL
Conjugate RY586
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD158b1/KIR2DL2/NKAT-6; CD158b2/KIR2DL3/NKAT-2; CD158j/KIR2DS2/NKAT-5
Host Species Mouse
Immunogen Human NK Cells
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.