Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD158i Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS2644R

Description
Killer cell immunoglobulin-like receptors (KIRs) are transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes (KIR3DL3, KIR3DP1, KIR3DL4, KIR3DL2). The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. The ligands for several KIR proteins are subsets of HLA class I molecules; thus, KIR proteins are thought to play an important role in regulation of the immune response.Specifications
CD158i | |
Polyclonal | |
Unconjugated | |
KIR2DS4 | |
CD158 antigen-like family member I; CD158I; killer cell immunoglobulin like receptor, two Ig domains and short cytoplasmic tail 4; Killer cell immunoglobulin-like receptor 2DS4; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 1; killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 4; killer Ig receptor; killer inhibitory receptor 4-1-2; killer-cell immunoglobulin-like receptor; KIR antigen 2DS4; KIR1D; KIR2DS1; KIR2DS4; KIR-2DS4; KIR412; KKA3; MHC class I NK cell receptor; natural killer cell inhibitory receptor; Natural killer-associated transcript 8; NKAT8; NKAT-8; p50 killer cell activating receptor KAR-K1d; P58 natural killer cell receptor clones CL-39/CL-17; p58 NK receptor CL-39/CL-17 | |
Rabbit | |
Protein A | |
RUO | |
3809 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P43632 | |
KIR2DS4 | |
KLH conjugated synthetic peptide derived from human CD158i . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction