Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD159a (NKG2AB6) Mouse anti-Mouse, APC, Clone: 16a11 (also known as 16A11), BD Phrmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569528
Description
CD159a (NKG2AB6) Monoclonal antibody specifically detects CD159a (NKG2AB6) in Mouse samples. It is validated for Flow cytometry
Specifications
CD159a (NKG2AB6) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
16641 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Flow Cytometry | |
16a11 (also known as 16A11) | |
APC | |
CD159a; NKG2A; NKG2A B6; NKG2A-B6 | |
C57BL/6 Mouse CD94:NKG2A transfected CHO cells | |
25 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction