Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/32 Rat anti-Mouse, APC, Clone: 93, Proteintech
SDP

Catalog No. 502256627 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6627 Supplier Proteintech Group Inc Supplier No. APC65057100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

The CD16/32 antibody from Proteintech is a rat monoclonal antibody generated with vh81x tg b cells and t220 stromal cell line of mouse CD16/32. This antibody recognizes mouse antigen. The CD16/32 antibody has been validated for the following applications: FC analysis. This antibody is apc.

Specifications

Antigen CD16/32
Applications Flow Cytometry
Classification Monoclonal
Clone 93
Concentration 0.1 mg/mL
Conjugate APC
Host Species Rat
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 14130
Target Species Mouse
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.