Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16/CD32 Antibody (93), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP127946F
Description
CD16/CD32 Monoclonal specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD16/CD32 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FCGR3A | |
Murine CD16/32 (CD16/Fc gamma III and CD32/FcgammaII receptors) | |
0.25 mL | |
Primary | |
Murine CD16/32 (CD16/Fc gamma III and CD32/Fc gamma II receptors) | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Frozen) | |
93 | |
Flow Cytometry, Immunohistochemistry-Frozen | |
CD16, CD16A, Fc fragment of IgG, low affinity IIIa, receptor (CD16a), FCG3, FCGR3, FCGR3A, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3 | |
Rat | |
Protein A or G purified | |
RUO | |
2214 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction