Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD16/CD32 Monoclonal Antibody (93), Biotin, eBioscience™, Invitrogen™

Catalog No. 5011449
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-114-49 500 μg
50-114-48 100 μg
50-114-50 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-114-49 Supplier Invitrogen™ Supplier No. 13016185
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils. Applications Reported: The 93 antibody has been reported for use in flow cytometric analysis. Applications Tested: The 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.

CD16 is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by natural killer (NK) cells, and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC (antibody dependent cell mediated cytotoxicity) by NK cells. IN comparison, CD32 is a 40 kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII), and mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Further, CD32 is involved in the phagocytosis of immune complexes, the regulation of antibody production by B-cells, and variations of the CD32 gene may increase vulnerability to systemic lupus erythematosus (SLE). Several transcript variants encoding different isoforms of CD32 have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16/CD32
Applications Flow Cytometry
Classification Monoclonal
Clone 93
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Fcgr3
Gene Accession No. P08101, P08508
Gene Alias AI528646; CD16; CD16a; CD16a antigen; CD16B; CD32; CD32 receptor 2; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIIa; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor (CD16a); fc gamma receptor IIB; Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor RII; Fc gamma RIIB; Fc gamma RIIIa; Fc receptor, IgG, low affinity IIb; Fc receptor, IgG, low affinity III; Fc[g]RII; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD16); fc-gamma RII; Fc-gamma RII-b; Fc-gamma RII-c; Fc-gamma RIII; Fc-gamma RIIIa; Fc-gamma RIII-alpha; Fc-gamma-RII; FcgammaRIIB; fc-gamma-RIIB; Fc-gamma-RIIc; FcgammaRIII; FcgammaRIIIA; Fcgr2; Fcgr2a; Fcgr2b; FCGR2C; Fcgr3; FCGR3A; FCGR3B; FcgRII; FCGRIII; FCR-10; Fcr-2; Fcr-3; fcRII; FcRII-b; FcRII-c; FCRIII; FCRIIIA; FGFR2B; IGFR3; IgG Fc receptor II beta; IgG Fc receptor III; IgG Fc receptor III-2; IMD20; immunoglobulin G Fc receptor III; low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; low affinity immunoglobulin gamma Fc region receptor III; Low affinity immunoglobulin gamma Fc region receptor III-A; Ly-17; Lym-1; Ly-m20; lymphocyte antigen 17; neutrophil-specific antigen NA; RP11-5K23.1
Gene Symbols Fcgr2b, Fcgr3
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14130, 14131
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.