Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Rat anti-Mouse, APC-R700, Clone: 2.4G2, BD Horizon™

Catalog No. BDB565502 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565502 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565502 Supplier BD Biosciences Supplier No. 565502
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16/CD32
Applications Flow Cytometry
Classification Monoclonal
Clone 2.4G2
Concentration 0.2mg/mL
Conjugate APC-R700
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias FcγRIII/FcγRII; Fcgr3/Fcgr2
Host Species Rat
Immunogen Mouse BALB/c Macrophage J774
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed.
Form Purified
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.