Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD16/CD32 Rat anti-Mouse, Brilliant Violet 786, Clone: 190909, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 747955
Description
CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.
Specifications
CD16/CD32 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
CD16/CD32 | |
Refrigerate (2 to 8°C) |
Flow Cytometry | |
190909 | |
Brilliant Violet 786 | |
50 μg | |
Primary | |
Mouse | |
IgG2b κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction