Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Rat anti-Mouse, BV711, Clone: Ab93 (also known as 93 or Antibody93), BD OptiBuild™

Catalog No. BDB751691 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751691 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751691 Supplier BD Biosciences Supplier No. 751691
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD16/CD32 Monoclonal antibody specifically detects CD16/CD32 in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD16/CD32
Applications Flow Cytometry
Classification Monoclonal
Clone Ab93 (also known as 93 or Antibody93)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 711
Description Ms CD16/CD32 BV711 AB93 50 μg
Host Species Rat
Immunogen Mouse Pre-B cells
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD16/CD32
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2a λ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.