Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD16/CD32 Monoclonal Antibody (93), Functional Grade, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

$111.00 - $648.00

Specifications

Antigen CD16/CD32
Clone 93
Concentration 1 mg/mL
Applications Flow Cytometry, Functional Assay, Neutralization
Classification Monoclonal
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-9580
Encompass_Preferred
View Documents
Invitrogen™
16016181
50 μg
Each for $111.00
Only null left
Edge
Add to Cart
 
50-142-41
GREENER_CHOICE
Encompass_Preferred
View Documents
Invitrogen™
16016182
100 μg
Each for $200.00
Only null left
Add to Cart
 
50-112-9629
Encompass_Preferred
View Documents
Invitrogen™
16016185
500 μg
Each for $389.00
Only null left
Add to Cart
 
50-112-9418
Encompass_Preferred
View Documents
Invitrogen™
16016186
1 mg
Each for $648.00
Only null left
Edge
Add to Cart
 
Description

Description

Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils. Applications Reported: The 93 antibody has been reported for use in flow cytometric analysis. 93 has also been reported in blocking of Fc-mediated reactions in functional studies. Applications Tested: The 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.

CD16 is a 50-65 kDa cell surface molecule that exists in two forms - a transmembranous form expressed by natural killer (NK) cells, and some T cells and a phosphatidylinositol linked form expressed by granulocytes. CD16 is a low affinity receptor for IgG (FcR III), and is an important receptor mediating ADCC (antibody dependent cell mediated cytotoxicity) by NK cells. IN comparison, CD32 is a 40 kD glycoprotein that acts as a low affinity receptor for IgG (also known as Fc gamma RII), and mediates several functions including endocytosis, activation of secretion, cytotoxicity and immunomodulation. CD32 is expressed by B cells, monocytes, granulocytes and platelets. Further, CD32 is involved in the phagocytosis of immune complexes, the regulation of antibody production by B-cells, and variations of the CD32 gene may increase vulnerability to systemic lupus erythematosus (SLE). Several transcript variants encoding different isoforms of CD32 have been found.
Specifications

Specifications

CD16/CD32
1 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.2
AI528646; CD16; CD16a; CD16a antigen; CD16B; CD32; CD32 receptor 2; CD32B; CDw32; F630109E10Rik; Fc fragment of IgG low affinity IIIa receptor; Fc fragment of IgG receptor IIb; Fc fragment of IgG receptor IIIa; Fc fragment of IgG, low affinity IIb, receptor (CD32); Fc fragment of IgG, low affinity III, receptor for (CD16); Fc fragment of IgG, low affinity IIIa, receptor (CD16a); fc gamma receptor IIB; Fc gamma receptor III; Fc gamma receptor IIIa; Fc gamma receptor III-A; Fc gamma receptor RII; Fc gamma RIIB; Fc gamma RIIIa; Fc receptor, IgG, low affinity IIb; Fc receptor, IgG, low affinity III; Fc[g]RII; Fcg receptor III; FCG2; FCG3; Fc-gamma receptor III-2 (CD 16); Fc-gamma receptor IIIb (CD16); fc-gamma RII; Fc-gamma RII-b; Fc-gamma RII-c; Fc-gamma RIII; Fc-gamma RIIIa; Fc-gamma RIII-alpha; Fc-gamma-RII; FcgammaRIIB; fc-gamma-RIIB; Fc-gamma-RIIc; FcgammaRIII; FcgammaRIIIA; Fcgr2; Fcgr2a; Fcgr2b; FCGR2C; Fcgr3; FCGR3A; FCGR3B; FcgRII; FCGRIII; FCR-10; Fcr-2; Fcr-3; fcRII; FcRII-b; FcRII-c; FCRIII; FCRIIIA; FGFR2B; IGFR3; IgG Fc receptor II beta; IgG Fc receptor III; IgG Fc receptor III-2; IMD20; immunoglobulin G Fc receptor III; low affinity immunoglobulin gamma Fc region receptor II; low affinity immunoglobulin gamma Fc region receptor II-b; low affinity immunoglobulin gamma Fc region receptor III; Low affinity immunoglobulin gamma Fc region receptor III-A; Ly-17; Lym-1; Ly-m20; lymphocyte antigen 17; neutrophil-specific antigen NA; RP11-5K23.1
Fcgr2b, Fcgr3
Primary
4°C
Fcgr3
93
Flow Cytometry, Functional Assay, Neutralization
Functional Grade
Rat
Mouse
P08101, P08508
14130, 14131
IgG2a λ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.