Learn More
Description
Host Species: Mouse
Clone: 2H7
Isotype: IgG2a
Species Reactivity: Human. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant protein encoding the external domain of human CD16
Molecular Weight: 50-65 kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin only)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD16 Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 2H7 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant protein encoding the external domain of human CD16 |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Signal Transduction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.